Assay Details
Target Gene Details
Entrez Gene ID: | 3189 |
Gene Name: | heterogeneous nuclear ribonucleoprotein H3 |
Gene Aliases: |
2H9, HNRPH3 |
Location: |
Chr.10:68331174-68343196 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HNRNPH3 | NM_001322434.1 | 1 | 138 | NP_001309363.1 |
| NM_001322436.1 | NP_001309365.1 | |||
| NM_001322438.1 | NP_001309367.1 | |||
| NM_001322439.1 | 1 | 138 | NP_001309368.1 | |
| NM_001322440.1 | NP_001309369.1 | |||
| NM_001322442.1 | 1 | 138 | NP_001309371.1 | |
| NM_001322443.1 | 1 | 138 | NP_001309372.1 | |
| NM_001322444.1 | 1 | 138 | NP_001309373.1 | |
| NM_001322445.1 | NP_001309374.1 | |||
| NM_001322446.1 | 1 | 138 | NP_001309375.1 | |
| NM_001322447.1 | NP_001309376.1 | |||
| NM_001322448.1 | 1 | 138 | NP_001309377.1 | |
| NM_001322450.1 | 1 | 138 | NP_001309379.1 | |
| NM_001322451.1 | NP_001309380.1 | |||
| NM_001322452.1 | 1 | 138 | NP_001309381.1 | |
| NM_001322453.1 | 1 | 138 | NP_001309382.1 | |
| NM_012207.2 | 1 | 138 | NP_036339.1 | |
| NM_021644.3 | 1 | 138 | NP_067676.2 | |
| XM_011539742.1 | 1 | 90 | XP_011538044.1 | |
| XM_017016168.1 | XP_016871657.1 | |||
| XM_017016169.1 | XP_016871658.1 | |||
| AF132361.1 | AAF68849.1 | |||
| AK091411.1 | 1 | 975 | ||
| AK223441.1 | 1 | 90 | BAD97161.1 | |
| AK291547.1 | 1 | 90 | ||
| AK295316.1 | 1 | 53 | ||
| BC039824.1 | 1 | 74 | AAH39824.1 | |
| DA486117.1 | 1 | 138 |
Target Gene Details
Entrez Gene ID: | 64081 |
Gene Name: | phenazine biosynthesis like protein domain containing |
Gene Aliases: |
HEL-S-306, MAWBP, MAWDBP |
Location: |
Chr.10:68282660-68332958 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PBLD | NM_001033083.1 | NP_001028255.1 | ||
| NM_022129.3 | NP_071412.2 | |||
| XM_005270028.3 | XP_005270085.1 | |||
| XM_011540060.2 | XP_011538362.1 | |||
| XM_017016513.1 | XP_016872002.1 | |||
| XM_017016514.1 | XP_016872003.1 | |||
| AK027673.1 | BAB55285.1 | |||
| BC009738.1 | AAH09738.1 | |||
| CB121561.1 | ||||
| GQ472233.1 |
More Information
Set Membership: |
|

Genomic Map