Assay Details
Target Gene Details
Entrez Gene ID: | 4686 |
Gene Name: | nuclear cap binding protein subunit 1 |
Gene Aliases: |
CBP80, NCBP, Sto1 |
Location: |
Chr.9:97633423-97673748 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCBP1 | NM_002486.4 | 1 | 177 | NP_002477.1 |
| XM_011518715.2 | 1 | 176 | XP_011517017.1 | |
| XM_017014735.1 | XP_016870224.1 | |||
| AK312807.1 | 1 | 177 | ||
| D32002.1 | BAA06769.1 | |||
| DB086376.1 | 1 | 177 |
Target Gene Details
Entrez Gene ID: | 158427 |
Gene Name: | thiosulfate sulfurtransferase like domain containing 2 |
Gene Aliases: |
C9orf97 |
Location: |
Chr.9:97600080-97633680 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSTD2 | NM_139246.4 | 1 | 82 | NP_640339.4 |
| DA735730.1 | 1 | 82 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1494e214 | Chr.9:97595020 - 97658603 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map