Assay Details
Target Gene Details
Entrez Gene ID: | 374395 |
Gene Name: | transmembrane protein 179B |
Gene Aliases: |
- |
Location: |
Chr.11:62787321-62790400 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM179B | NM_199337.2 | NP_955369.1 | ||
| XM_005273982.3 | XP_005274039.3 | |||
| BC051355.1 | AAH51355.1 | |||
| BE439533.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508636 | Chr.11:62779642 - 62876986 on Build GRCh38 | Deletion |
|

Genomic Map