Assay Details
Target Gene Details
Entrez Gene ID: | 51102 |
Gene Name: | mitochondrial trans-2-enoyl-CoA reductase |
Gene Aliases: |
CGI-63, FASN2B, NRBF1 |
Location: |
Chr.1:29192657-29230958 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
More Information
Set Membership: |
|

Genomic Map