Assay Details
Target Gene Details
Entrez Gene ID: | 10946 |
Gene Name: | splicing factor 3a subunit 3 |
Gene Aliases: |
PRP9, PRPF9, SAP61, SF3a60 |
Location: |
Chr.1:37956975-37990110 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SF3A3 | NM_001320830.1 | 1 | 123 | NP_001307759.1 |
| NM_006802.3 | 1 | 123 | NP_006793.1 | |
| AK128438.1 | 1 | 934 | ||
| AK222554.1 | BAD96274.1 | |||
| AK291181.1 | ||||
| AK292116.1 | ||||
| AK301424.1 | ||||
| BC002395.2 | AAH02395.1 | |||
| BC011523.1 | 1 | 80 | AAH11523.1 | |
| DA835019.1 | 1 | 123 | ||
| U08815.1 | AAA19625.1 |
More Information
Set Membership: |
|

Genomic Map