Assay Details
Target Gene Details
Entrez Gene ID: | 27198 |
Gene Name: | hydroxycarboxylic acid receptor 1 |
Gene Aliases: |
FKSG80, GPR104, GPR81, HCA1, LACR1, TA-GPCR, TAGPCR |
Location: |
Chr.12:122727606-122730582 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HCAR1 | NM_032554.3 | 1 | 1490 | NP_115943.1 |
| AF345568.1 | AAK29071.1 | |||
| AF385431.1 | 1 | 1490 | AAM43832.1 | |
| AF385432.1 | 1 | 1490 | AAM43833.1 | |
| AK122659.1 | ||||
| AK289557.1 | 1 | 1674 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv913 | Chr.12:122705485 - 122732302 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map