Assay Details
Target Gene Details
Entrez Gene ID: | 1773 |
Gene Name: | deoxyribonuclease I |
Gene Aliases: |
DNL1, DRNI |
Location: |
Chr.16:3611705-3665472 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNASE1 | NM_005223.3 | 9 | 3042 | NP_005214.2 |
| XM_005255148.3 | XP_005255205.1 | |||
| XM_006720854.3 | 11 | 1865 | XP_006720917.1 | |
| XM_011522393.2 | 12 | 1952 | XP_011520695.1 | |
| XM_017022992.1 | XP_016878481.1 | |||
| XM_017022993.1 | XP_016878482.1 | |||
| XM_017022994.1 | XP_016878483.1 | |||
| XM_017022995.1 | XP_016878484.1 | |||
| XM_017022996.1 | XP_016878485.1 | |||
| XM_017022997.1 | XP_016878486.1 | |||
| XM_017022998.1 | XP_016878487.1 | |||
| XM_017022999.1 | 10 | 1696 | XP_016878488.1 | |
| XM_017023000.1 | XP_016878489.1 | |||
| XM_017023001.1 | XP_016878490.1 | |||
| XM_017023002.1 | XP_016878491.1 | |||
| XM_017023003.1 | XP_016878492.1 | |||
| XM_017023004.1 | XP_016878493.1 | |||
| XM_017023005.1 | XP_016878494.1 | |||
| XM_017023006.1 | XP_016878495.1 | |||
| XM_017023007.1 | 10 | 1929 | XP_016878496.1 | |
| AB188151.1 | 10 | 1121 | ||
| AB188152.1 | 9 | 1175 | ||
| AI378682.1 | 1 | 70 | ||
| AJ298844.1 | 9 | 1997 | CAC12813.1 | |
| AK300914.1 | ||||
| BC029437.1 | 9 | 1063 | AAH29437.1 | |
| BX648257.1 | 6 | 1040 |
Target Gene Details
Entrez Gene ID: | 10131 |
Gene Name: | TNF receptor associated protein 1 |
Gene Aliases: |
HSP 75, HSP75, HSP90L, TRAP-1 |
Location: |
Chr.16:3658037-3717597 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRAP1 | NM_001272049.1 | NP_001258978.1 | ||
| NM_016292.2 | NP_057376.2 | |||
| XM_011522345.1 | XP_011520647.1 | |||
| XM_017022851.1 | XP_016878340.1 | |||
| AB209805.1 | BAD93042.1 | |||
| AF043254.1 | AAC02679.1 | |||
| AF154108.1 | AAF15314.1 | |||
| AJ890085.1 | CAI64498.1 | |||
| AK057620.1 | ||||
| AK093344.1 | BAC04139.1 | |||
| AK223049.1 | BAD96769.1 | |||
| AK223076.1 | BAD96796.1 | |||
| AK223205.1 | BAD96925.1 | |||
| AK299127.1 | ||||
| BC001455.2 | AAH01455.2 | |||
| BC018950.2 | AAH18950.1 | |||
| BC023585.2 | AAH23585.1 | |||
| U12595.1 | AAA87704.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833128 | Chr.16:3525122 - 3690497 on Build GRCh38 | Loss |
|
| esv2761892 | Chr.16:3651576 - 3669343 on Build GRCh38 | Loss |
|
| nsv518509 | Chr.16:3583656 - 3688960 on Build GRCh38 | Gain |
|
| nsv525301 | Chr.16:3657746 - 3668847 on Build GRCh38 | Loss |
|
| esv3637673 | Chr.16:3653616 - 3669650 on Build GRCh38 | Loss |
|
| nsv1051657 | Chr.16:3644759 - 3788297 on Build GRCh38 | Gain |
|
| nsv457359 | Chr.16:3655014 - 3668847 on Build GRCh38 | Loss |
|
| esv2659167 | Chr.16:3653585 - 3669799 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map