Assay Details
Target Gene Details
Entrez Gene ID: | 8707 |
Gene Name: | beta-1,3-galactosyltransferase 2 |
Gene Aliases: |
BETA3GALT2, GLCT2, beta3Gal-T2 |
Location: |
Chr.1:193178730-193186613 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B3GALT2 | NM_003783.3 | 2 | 789 | NP_003774.1 |
| AF288390.1 | 2 | 770 | AAG60610.1 | |
| AK314116.1 | 2 | 507 | ||
| BC022507.1 | 2 | 789 | AAH22507.1 | |
| Y15060.1 | 1 | 74 | CAA75344.1 |
Target Gene Details
Entrez Gene ID: | 79577 |
Gene Name: | cell division cycle 73 |
Gene Aliases: |
C1orf28, FIHP, HPTJT, HRPT1, HRPT2, HYX |
Location: |
Chr.1:193121958-193254815 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDC73 | NM_024529.4 | NP_078805.3 | ||
| AF312865.1 | AAG45339.1 | |||
| AK026969.1 | BAB15608.1 | |||
| AK226038.1 | ||||
| AK300929.1 | ||||
| AK314772.1 | ||||
| BC014351.1 | AAH14351.2 | |||
| BC065037.1 | AAH65037.1 | |||
| KU178771.1 | ||||
| KU178772.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832148 | Chr.1:193174815 - 193323767 on Build GRCh38 | Gain |
|
| nsv466994 | Chr.1:192394185 - 193552205 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map