Assay Details
Target Gene Details
Entrez Gene ID: | 3766 |
Gene Name: | potassium voltage-gated channel subfamily J member 10 |
Gene Aliases: |
BIRK-10, KCNJ13-PEN, KIR1.2, KIR4.1, SESAME |
Location: |
Chr.1:160037467-160070261 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNJ10 | NM_002241.4 | 2 | 2552 | NP_002232.2 |
| BC034036.1 | 2 | 2454 | AAH34036.2 | |
| BC131627.1 | 2 | 2448 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3199 | Chr.1:160033106 - 160064505 on Build GRCh38 | Insertion |
|
| esv2762195 | Chr.1:160030645 - 160040361 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map