Assay Details
Target Gene Details
Entrez Gene ID: | 56751 |
Gene Name: | BarH like homeobox 1 |
Gene Aliases: |
- |
Location: |
Chr.9:132582606-132590253 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BARHL1 | NM_020064.3 | NP_064448.1 | ||
| AF321618.1 | AAK52674.1 | |||
| AF325688.1 | AAG50279.1 | |||
| AK054797.1 | BAB70807.1 | |||
| BC136956.1 | ||||
| BC136969.1 | ||||
| BC144433.1 |
More Information
Set Membership: |
|

Genomic Map