Assay Details
Target Gene Details
Entrez Gene ID: | 8824 |
Gene Name: | carboxylesterase 2 |
Gene Aliases: |
CE-2, CES2A1, PCE-2, iCE |
Location: |
Chr.16:66934444-66945096 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CES2 | NM_003869.5 | 1 | 182 | NP_003860.2 |
| NM_198061.2 | 1 | 182 | NP_932327.1 | |
| NR_036684.1 | 1 | 155 | ||
| AK095522.1 | 1 | 155 | ||
| AL713761.1 | 1 | 135 | CAD28531.1 | |
| BC032095.2 | 1 | 166 | AAH32095.1 | |
| BX538086.1 | 1 | 183 | CAD98009.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv827706 | Chr.16:66894354 - 67009614 on Build GRCh38 | Loss |
|
| nsv572904 | Chr.16:66876182 - 66938470 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map