Assay Details
Target Gene Details
Entrez Gene ID: | 112476 |
Gene Name: | proline rich transmembrane protein 2 |
Gene Aliases: |
BFIC2, BFIS2, DSPB3, DYT10, EKD1, FICCA, ICCA, IFITMD1, PKC |
Location: |
Chr.16:29811856-29815920 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRRT2 | NM_001256442.1 | 2 | 762 | NP_001243371.1 |
| NM_001256443.1 | 2 | 790 | NP_001243372.1 | |
| NM_145239.2 | 2 | 762 | NP_660282.2 | |
| XM_011545715.2 | 2 | 691 | XP_011544017.1 | |
| XM_011545716.2 | 2 | 707 | XP_011544018.1 | |
| XM_017022887.1 | 2 | 757 | XP_016878376.1 | |
| XM_017022888.1 | 2 | 691 | XP_016878377.1 | |
| XM_017022889.1 | 2 | 757 | XP_016878378.1 | |
| AK074572.1 | 2 | 656 | BAC11067.1 | |
| AK092265.1 | 2 | 635 | BAC03843.1 | |
| AK098379.1 | 3 | 474 | ||
| AK225785.1 | 2 | 656 | ||
| AK292393.1 | 2 | 659 | ||
| AL834185.1 | 2 | 601 | CAD38881.1 | |
| BC011405.1 | 2 | 652 | AAH11405.1 | |
| BC053594.1 | 2 | 581 | AAH53594.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953800 | Chr.16:29804780 - 29816179 on Build GRCh38 | Deletion |
|
| nsv833186 | Chr.16:29719552 - 29863674 on Build GRCh38 | Loss |
|
| nsv827610 | Chr.16:29808186 - 29818225 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map