Assay Details
Target Gene Details
Entrez Gene ID: | 3700 |
Gene Name: | inter-alpha-trypsin inhibitor heavy chain family member 4 |
Gene Aliases: |
GP120, H4P, IHRP, ITI-HC4, ITIHL1, PK-120, PK120 |
Location: |
Chr.3:52812990-52830701 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITIH4 | NM_001166449.1 | NP_001159921.1 | ||
| NM_002218.4 | NP_002209.2 | |||
| AB209388.1 | BAD92625.1 | |||
| AK298407.1 | ||||
| AK298412.1 | ||||
| AK298429.1 | ||||
| AK303769.1 | ||||
| BC136392.1 | ||||
| BC136393.1 | ||||
| BX647322.1 | ||||
| D38595.1 | BAA07602.1 |
Target Gene Details
Entrez Gene ID: | 100873993 |
Gene Name: | ITIH4 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:52823935-52825314 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITIH4-AS1 | NR_046615.1 | |||
| AI004860.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv590308 | Chr.3:52820319 - 52826790 on Build GRCh38 | Loss |
|
| nsv508928 | Chr.3:52769883 - 52888575 on Build GRCh38 | Insertion |
|
| esv2762328 | Chr.3:52740868 - 52968134 on Build GRCh38 | Loss |
|
| nsv834698 | Chr.3:52715967 - 52894203 on Build GRCh38 | Gain |
|
| nsv3822 | Chr.3:52799815 - 52832115 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map