Assay Details
Target Gene Details
Entrez Gene ID: | 1305 |
Gene Name: | collagen type XIII alpha 1 chain |
Gene Aliases: |
CMS19, COLXIIIA1 |
Location: |
Chr.10:69801885-69959148 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv32737 | Chr.10:69504230 - 70786847 on Build GRCh38 | Gain+Loss |
|
| nsv428233 | Chr.10:69681990 - 70004998 on Build GRCh38 | Gain |
|
| esv2761603 | Chr.10:69790657 - 69824001 on Build GRCh38 | Gain |
|
| nsv516370 | Chr.10:69801243 - 69820364 on Build GRCh38 | Gain |
|
| nsv551478 | Chr.10:69798188 - 69822699 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map