Assay Details
Target Gene Details
Entrez Gene ID: | 165721 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member B8 |
Gene Aliases: |
CT156, DJ6 |
Location: |
Chr.3:128462432-128467248 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJB8 | NM_153330.4 | NP_699161.1 | ||
| XM_006713519.3 | 1 | 328 | XP_006713582.1 | |
| AK125946.1 | 1 | 328 | ||
| BC050288.1 | AAH50288.1 |
Target Gene Details
Entrez Gene ID: | 285224 |
Gene Name: | DNAJB8 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:128463594-128472317 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJB8-AS1 | NR_037890.1 | |||
| AK097457.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759176 | Chr.3:128338850 - 128718974 on Build GRCh38 | Loss |
|
| nsv508240 | Chr.3:128347675 - 128482477 on Build GRCh38 | Deletion |
|
| nsv470905 | Chr.3:128451019 - 128473810 on Build GRCh38 | Loss |
|
| nsv516926 | Chr.3:128378653 - 128503934 on Build GRCh38 | Loss |
|
| dgv8565n54 | Chr.3:128403977 - 128469086 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map