Assay Details
Target Gene Details
Entrez Gene ID: | 808 |
Gene Name: | calmodulin 3 |
Gene Aliases: |
HEL-S-72, PHKD, PHKD3 |
Location: |
Chr.19:46601255-46610782 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CALM3 | NM_005184.2 | NP_005175.2 | ||
| AK094964.1 | 2 | 467 | ||
| AK295927.1 | ||||
| AK310762.1 | ||||
| BC005137.2 | AAH05137.1 | |||
| BC006182.1 | AAH06182.1 | |||
| BG753029.1 | ||||
| BT006855.1 | AAP35501.1 | |||
| FJ224310.1 | ||||
| J04046.1 | AAA51918.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3352253 | Chr.19:46605755 - 46608653 on Build GRCh38 | Duplication |
|
| nsv833849 | Chr.19:46558831 - 46703060 on Build GRCh38 | Loss |
|
| nsv953589 | Chr.19:46601544 - 46636643 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map