Assay Details
Target Gene Details
Entrez Gene ID: | 6643 |
Gene Name: | sorting nexin 2 |
Gene Aliases: |
TRG-9 |
Location: |
Chr.5:122774996-122834539 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNX2 | NM_001278199.1 | 1 | 95 | NP_001265128.1 |
| NM_003100.3 | 1 | 95 | NP_003091.2 | |
| AF043453.1 | AAB99852.1 | |||
| AF065482.1 | AAC17181.1 | |||
| AK002170.1 | ||||
| AK023581.1 | ||||
| AK023716.1 | ||||
| AK222968.1 | BAD96688.1 | |||
| AY272044.1 | AAQ02693.1 | |||
| BC003382.1 | AAH03382.1 | |||
| BT009841.1 | AAP88843.1 | |||
| DC313082.1 | 1 | 95 | ||
| DC338761.1 | 1 | 139 |
More Information
Set Membership: |
|

Genomic Map