Assay Details
Target Gene Details
Entrez Gene ID: | 112936 |
Gene Name: | VPS26, retromer complex component B |
Gene Aliases: |
Pep8b |
Location: |
Chr.11:134224605-134247792 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VPS26B | NM_052875.4 | NP_443107.1 | ||
| XM_011542565.2 | XP_011540867.1 | |||
| AF452718.1 | AAP13353.1 | |||
| BC009747.2 | AAH09747.1 | |||
| BC014128.2 | AAH14128.1 | |||
| BC098386.1 | AAH98386.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521581 | Chr.11:134126584 - 134772738 on Build GRCh38 | Gain |
|
| esv3892114 | Chr.11:134054794 - 134328278 on Build GRCh38 | Gain |
|
| nsv832299 | Chr.11:134170258 - 134346785 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map