Assay Details
Target Gene Details
Entrez Gene ID: | 6508 |
Gene Name: | solute carrier family 4 member 3 |
Gene Aliases: |
AE3, CAE3/BAE3, SLC2C |
Location: |
Chr.2:219627570-219641980 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC4A3 | NM_001326559.1 | 3 | 376 | NP_001313488.1 |
| NM_005070.3 | 3 | 376 | NP_005061.2 | |
| NM_201574.2 | 3 | 316 | NP_963868.2 | |
| NR_048551.1 | 3 | 376 | ||
| XM_005246789.4 | 3 | 316 | XP_005246846.1 | |
| XM_011511665.2 | 3 | 311 | XP_011509967.1 | |
| AK226122.1 | 3 | 246 | ||
| AK289974.1 | 3 | 310 | ||
| AK299109.1 | 2 | 142 | ||
| AK309025.1 | 3 | 316 | ||
| AK314083.1 | 3 | 320 | ||
| AY142112.1 | 3 | 326 | AAN34939.1 | |
| BC136384.1 | 2 | 143 | ||
| BC146656.1 | 3 | 244 | ||
| BC171760.1 | 2 | 143 | ||
| U05596.1 | 2 | 118 | AAA50748.1 |
More Information
Set Membership: |
|

Genomic Map