Assay Details
Target Gene Details
Entrez Gene ID: | 222171 |
Gene Name: | proline rich 15 |
Gene Aliases: |
- |
Location: |
Chr.7:29563796-29567300 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRR15 | NM_175887.2 | 2 | 1288 | NP_787083.1 |
| XM_005249653.3 | 2 | 2091 | XP_005249710.1 | |
| XM_011515198.1 | 2 | 2086 | XP_011513500.1 | |
| XM_011515199.1 | 2 | 1000 | XP_011513501.1 | |
| XM_017011842.1 | 2 | 1298 | XP_016867331.1 | |
| BC029131.1 | 2 | 1288 | AAH29131.1 | |
| FJ857942.1 | 1 | 608 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830935 | Chr.7:29530312 - 29753174 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map