Assay Details
Target Gene Details
Entrez Gene ID: | 24138 |
Gene Name: | interferon induced protein with tetratricopeptide repeats 5 |
Gene Aliases: |
ISG58, P58, RI58 |
Location: |
Chr.10:89414568-89421002 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IFIT5 | NM_012420.2 | 2 | 954 | NP_036552.1 |
| AK293346.1 | 3 | 616 | ||
| AK312358.1 | 2 | 787 | ||
| BC025786.1 | 2 | 933 | AAH25786.1 | |
| CR457031.1 | 1 | 723 | CAG33312.1 | |
| U34605.1 | 2 | 840 | AAA84934.1 |
More Information
Set Membership: |
|

Genomic Map