Assay Details
Target Gene Details
Entrez Gene ID: | 246176 |
Gene Name: | growth arrest specific 2 like 2 |
Gene Aliases: |
GAR17 |
Location: |
Chr.17:35744511-35752878 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GAS2L2 | NM_139285.3 | 6 | 1587 | NP_644814.1 |
| AF508784.1 | 7 | 1540 | AAM34264.1 | |
| AF508785.1 | 6 | 1587 | AAM34265.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv1004453 | Chr.17:35744536 - 35753153 on Build GRCh38 | Deletion |
|
| nsv517530 | Chr.17:35745536 - 35750409 on Build GRCh38 | Loss |
|
| esv3892999 | Chr.17:35252863 - 35925533 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map