Assay Details
Target Gene Details
Entrez Gene ID: | 369 |
Gene Name: | A-Raf proto-oncogene, serine/threonine kinase |
Gene Aliases: |
A-RAF, ARAF1, PKS2, RAFA1 |
Location: |
Chr.X:47561100-47571921 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARAF | NM_001256196.1 | 7 | 857 | NP_001243125.1 |
| NM_001654.4 | 7 | 848 | NP_001645.1 | |
| XM_006724529.2 | 7 | 784 | XP_006724592.1 | |
| XM_011543906.2 | 7 | 2174 | XP_011542208.1 | |
| XM_011543908.2 | 7 | 2158 | XP_011542210.1 | |
| XM_011543909.2 | 2 | 149 | XP_011542211.1 | |
| AB208831.1 | 6 | 2033 | BAD92068.1 | |
| AK130043.1 | 7 | 741 | ||
| AK297453.1 | ||||
| AK300975.1 | 7 | 704 | ||
| AK307747.1 | 7 | 741 | ||
| BC002466.2 | 7 | 728 | AAH02466.1 | |
| BC007514.2 | 7 | 735 | AAH07514.1 | |
| BT019864.1 | 6 | 637 | AAV38667.1 | |
| KU177914.1 | 6 | 637 | ||
| KU177915.1 | 6 | 652 | ||
| X04790.1 | 7 | 831 | CAA28476.1 |
More Information
Set Membership: |
|

Genomic Map