Assay Details
Target Gene Details
Entrez Gene ID: | 27289 |
Gene Name: | Rho family GTPase 1 |
Gene Aliases: |
ARHS, RHO6, RHOS |
Location: |
Chr.12:48857133-48865870 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RND1 | NM_014470.3 | 5 | 892 | NP_055285.1 |
| AB040147.1 | 5 | 870 | BAB17851.1 | |
| AK124288.1 | 1 | 1377 | ||
| AK292762.1 | 5 | 892 | ||
| AL359573.1 | 1 | 125 | ||
| BC026356.1 | 5 | 892 | AAH26356.1 |
More Information
Set Membership: |
|

Genomic Map