Assay Details
Target Gene Details
Entrez Gene ID: | 58487 |
Gene Name: | CREB/ATF bZIP transcription factor |
Gene Aliases: |
SMILE, ZF |
Location: |
Chr.11:85657564-85665148 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CREBZF | NM_001039618.2 | 1 | 5757 | NP_001034707.1 |
| NR_028024.1 | 2 | 4691 | ||
| NR_028025.1 | 3 | 4478 | ||
| NR_028026.1 | 5 | 2800 | ||
| NR_028027.1 | 4 | 2687 | ||
| XM_006718642.3 | 3 | 5324 | XP_006718705.1 | |
| XM_011545195.2 | 2 | 5537 | XP_011543497.1 | |
| XM_017018086.1 | 4 | 4603 | XP_016873575.1 | |
| XM_017018087.1 | 5 | 2812 | XP_016873576.1 | |
| XM_017018088.1 | 2 | 5046 | XP_016873577.1 | |
| XM_017018089.1 | 2 | 3976 | XP_016873578.1 | |
| XM_017018090.1 | 3 | 4816 | XP_016873579.1 | |
| XM_017018091.1 | 3 | 2910 | XP_016873580.1 | |
| XM_017018092.1 | 4 | 3025 | XP_016873581.1 | |
| AL110236.1 | 1 | 693 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759845 | Chr.11:85469436 - 85892945 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map