Assay Details
Target Gene Details
Entrez Gene ID: | 81285 |
Gene Name: | olfactory receptor family 51 subfamily E member 2 |
Gene Aliases: |
OR51E3P, OR52A2, PSGR |
Location: |
Chr.11:4680171-4697846 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR51E2 | NM_030774.3 | 2 | 1094 | NP_110401.1 |
| AF369708.1 | 2 | 1000 | AAK38728.1 | |
| AK314051.1 | 2 | 1094 | ||
| AY033942.1 | 2 | 1091 | AAK57550.1 | |
| BC020768.1 | 3 | 967 | AAH20768.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891914 | Chr.11:4642313 - 4974456 on Build GRCh38 | Gain |
|
| nsv1054847 | Chr.11:4621645 - 5179426 on Build GRCh38 | Gain |
|
| nsv832056 | Chr.11:4651771 - 4803660 on Build GRCh38 | Gain |
|
| nsv832055 | Chr.11:4529664 - 4715110 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map