Assay Details
Target Gene Details
Entrez Gene ID: | 124221 |
Gene Name: | protease, serine, 30 pseudogene |
Gene Aliases: |
Disp, TMPRSS8, TMPRSS8P |
Location: |
Chr.16:2839573-2842751 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRSS30P | NR_026864.2 | 1 | 1411 | |
| AK122946.1 | 1 | 1298 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952905 | Chr.16:2817200 - 2863199 on Build GRCh38 | Deletion |
|

Genomic Map