Assay Details
Target Gene Details
Entrez Gene ID: | 2823 |
Gene Name: | glycoprotein M6A |
Gene Aliases: |
GPM6, M6A |
Location: |
Chr.4:175632937-176002691 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPM6A | NM_001261447.1 | NP_001248376.1 | ||
| NM_005277.4 | NP_005268.1 | |||
| NM_201591.2 | NP_963885.1 | |||
| NM_201592.2 | NP_963886.1 | |||
| NR_048571.1 | ||||
| XM_011531877.1 | XP_011530179.1 | |||
| AK226176.1 | ||||
| AX772804.1 | CAE06251.1 | |||
| BC022508.1 | AAH22508.1 | |||
| BC022528.1 | AAH22528.1 | |||
| BC044612.1 | AAH44612.1 | |||
| BI544825.1 | ||||
| BI546539.1 | ||||
| BI596896.1 | ||||
| BI670059.1 | ||||
| D49958.1 | BAA08712.1 | |||
| DA190376.1 | ||||
| DA507772.1 | ||||
| DA528125.1 | ||||
| DA621566.1 | ||||
| DA915764.1 | ||||
| H16369.1 |
Target Gene Details
Entrez Gene ID: | 101928590 |
Gene Name: | uncharacterized LOC101928590 |
Gene Aliases: |
- |
Location: |
Chr.4:175790307-175812189 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928590 | NR_125901.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3603269 | Chr.4:175788318 - 175889410 on Build GRCh38 | Gain |
|
| esv3603268 | Chr.4:175782989 - 175950469 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map