Assay Details
Target Gene Details
Entrez Gene ID: | 7014 |
Gene Name: | telomeric repeat binding factor 2 |
Gene Aliases: |
TRBF2, TRF2 |
Location: |
Chr.16:69355561-69386004 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TERF2 | NM_005652.4 | 10 | 2514 | NP_005643.2 |
| XM_005256121.3 | 11 | 2539 | XP_005256178.1 | |
| XM_005256122.3 | 11 | 2536 | XP_005256179.1 | |
| XM_005256123.3 | 10 | 2527 | XP_005256180.1 | |
| XM_005256124.3 | 8 | 2296 | XP_005256181.1 | |
| AF002999.1 | 10 | 2495 | AAB81135.1 | |
| BX090758.1 | 1 | 136 |
More Information
Additional Information:
For this assay, SNP(s) [rs114396464] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map