Assay Details
Target Gene Details
Entrez Gene ID: | 57522 |
Gene Name: | SLIT-ROBO Rho GTPase activating protein 1 |
Gene Aliases: |
ARHGAP13, NMTC2 |
Location: |
Chr.12:63844372-64147863 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRGAP1 | NM_020762.2 | NP_065813.1 | ||
| XM_005269042.3 | 1 | 393 | XP_005269099.1 | |
| XM_011538580.1 | XP_011536882.1 | |||
| BC053903.1 | AAH53903.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2751110 | Chr.12:63545953 - 63880900 on Build GRCh38 | Gain |
|
| esv2759905 | Chr.12:63508021 - 63858392 on Build GRCh38 | Gain |
|
| nsv832442 | Chr.12:63834291 - 64048991 on Build GRCh38 | Gain |
|
| nsv832441 | Chr.12:63669996 - 63858392 on Build GRCh38 | Loss |
|
| nsv1046836 | Chr.12:62998611 - 63983389 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map