Assay Details
Target Gene Details
Entrez Gene ID: | 60675 |
Gene Name: | prokineticin 2 |
Gene Aliases: |
BV8, HH4, KAL4, MIT1, PK2 |
Location: |
Chr.3:71771655-71785206 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PROK2 | NM_001126128.1 | 2 | 287 | NP_001119600.1 |
| NM_021935.3 | 2 | 287 | NP_068754.1 | |
| XM_017006974.1 | 2 | 221 | XP_016862463.1 | |
| AF182069.2 | 2 | 118 | AAG16893.2 | |
| AF333025.1 | 2 | 142 | AAK49919.1 | |
| AY349131.1 | 2 | 133 | AAR06657.1 | |
| BC069395.1 | 2 | 142 | AAH69395.1 | |
| BC096695.1 | 2 | 142 | AAH96695.1 | |
| BC098110.1 | 2 | 142 | AAH98110.1 | |
| BC098162.1 | 2 | 142 | AAH98162.1 |
More Information
Set Membership: |
|

Genomic Map