Assay Details
Target Gene Details
Entrez Gene ID: | 4878 |
Gene Name: | natriuretic peptide A |
Gene Aliases: |
ANF, ANP, ATFB6, ATRST2, CDD, CDD-ANF, CDP, PND |
Location: |
Chr.1:11845709-11847783 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPPA | NM_006172.3 | NP_006163.1 | ||
| BC005893.1 | AAH05893.1 | |||
| M30262.1 | AAA35669.1 |
Target Gene Details
Entrez Gene ID: | 100379251 |
Gene Name: | NPPA antisense RNA 1 |
Gene Aliases: |
NPPA-AS, NPPAAS |
Location: |
Chr.1:11840319-11847616 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPPA-AS1 | NR_037806.1 | |||
| BQ776822.1 | ||||
| CA773133.1 | ||||
| FJ706070.1 | ||||
| FJ706071.1 | ||||
| FJ706072.1 | ||||
| FJ706073.1 | ||||
| FJ706074.1 | ||||
| FJ706075.1 | ||||
| FJ706076.1 | ||||
| FJ706077.1 | ||||
| FJ706078.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv527521 | Chr.1:11840768 - 11849457 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72639211] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map