Assay Details
Target Gene Details
Entrez Gene ID: | 1376 |
Gene Name: | carnitine palmitoyltransferase 2 |
Gene Aliases: |
CPT1, CPTASE, IIAE4 |
Location: |
Chr.1:53196429-53214197 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPT2 | NM_000098.2 | 4 | 1093 | NP_000089.1 |
| XM_005270484.1 | 4 | 761 | XP_005270541.1 | |
| AK223052.1 | 2 | 336 | BAD96772.1 | |
| AK312687.1 | 4 | 697 | ||
| BC002445.2 | 4 | 702 | AAH02445.1 | |
| BC005172.2 | 4 | 702 | AAH05172.1 | |
| M58581.1 | 4 | 679 | AAB59462.1 | |
| U09648.1 | 4 | 1094 | AAB60383.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829871 | Chr.1:53200871 - 53383048 on Build GRCh38 | Loss |
|
| esv2422213 | Chr.1:53179891 - 53258767 on Build GRCh38 | Duplication |
|
| nsv1001535 | Chr.1:52620368 - 53346539 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map