Assay Details
Target Gene Details
Entrez Gene ID: | 200634 |
Gene Name: | keratinocyte associated protein 3 |
Gene Aliases: |
KCP3 |
Location: |
Chr.2:27442366-27446481 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRTCAP3 | NM_001168364.1 | NP_001161836.1 | ||
| NM_001321325.1 | NP_001308254.1 | |||
| NM_173853.3 | NP_776252.2 | |||
| AY157576.1 | AAO13160.1 | |||
| AY358993.1 | AAQ89352.1 | |||
| BC101687.1 | AAI01688.1 | |||
| BC101689.1 | AAI01690.1 | |||
| BC143574.1 | ||||
| BM794111.1 | ||||
| CB160871.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833614 | Chr.2:27263559 - 27452213 on Build GRCh38 | Gain |
|
| nsv817684 | Chr.2:27434300 - 27623736 on Build GRCh38 | Gain |
|
| nsv581244 | Chr.2:27335385 - 27480628 on Build GRCh38 | Loss |
|

Genomic Map