Assay Details
Target Gene Details
Entrez Gene ID: | 90313 |
Gene Name: | tumor protein p53 inducible protein 13 |
Gene Aliases: |
DSCP1 |
Location: |
Chr.17:29568067-29573157 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TP53I13 | NM_138349.2 | NP_612358.3 | ||
| XM_017025282.1 | XP_016880771.1 | |||
| AB110209.1 | BAD08203.1 | |||
| AK075341.1 | BAC11555.1 | |||
| BC001593.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521557 | Chr.17:29562968 - 29595257 on Build GRCh38 | Loss |
|
| nsv833409 | Chr.17:29465646 - 29655807 on Build GRCh38 | Gain |
|

Genomic Map