Assay Details
Target Gene Details
Entrez Gene ID: | 337968 |
Gene Name: | keratin associated protein 6-3 |
Gene Aliases: |
KAP6.3 |
Location: |
Chr.21:30592440-30593055 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRTAP6-3 | NM_181605.3 | 1 | 399 | NP_853636.3 |
| AB096951.1 | 1 | 399 | BAE46366.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762105 | Chr.21:30547730 - 30624847 on Build GRCh38 | Gain |
|
| nsv1057467 | Chr.21:30318003 - 30993921 on Build GRCh38 | Gain |
|
| nsv587354 | Chr.21:29978351 - 30620545 on Build GRCh38 | Gain |
|

Genomic Map