Assay Details
Target Gene Details
Entrez Gene ID: | 873 |
Gene Name: | carbonyl reductase 1 |
Gene Aliases: |
CBR, SDR21C1, hCBR1 |
Location: |
Chr.21:36069924-36073177 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CBR1 | NM_001286789.1 | 3 | 1257 | NP_001273718.1 |
| NM_001757.3 | 3 | 626 | NP_001748.1 | |
| AB124848.1 | 3 | 626 | BAE45940.1 | |
| AK311219.1 | 3 | 1157 | ||
| AK314879.1 | 3 | 524 | ||
| AL548852.3 | 3 | 563 | ||
| BC002511.2 | 3 | 511 | AAH02511.1 | |
| BC015640.2 | 3 | 525 | AAH15640.1 | |
| BT019843.1 | 3 | 434 | AAV38646.1 | |
| CR541708.1 | 3 | 434 | CAG46509.1 | |
| J04056.1 | 3 | 527 | AAA52070.1 |
Target Gene Details
Entrez Gene ID: | 100133286 |
Gene Name: | uncharacterized LOC100133286 |
Gene Aliases: |
- |
Location: |
Chr.21:36069642-36126640 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100133286 | NR_040084.1 | |||
| AK303953.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2751927 | Chr.21:36061467 - 36286211 on Build GRCh38 | Gain |
|
| nsv587461 | Chr.21:36003345 - 36087866 on Build GRCh38 | Loss |
|
| nsv470899 | Chr.21:36001424 - 36096978 on Build GRCh38 | Loss |
|
| nsv964547 | Chr.21:36003423 - 36090261 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map