Assay Details
Target Gene Details
Entrez Gene ID: | 140459 |
Gene Name: | ankyrin repeat and SOCS box containing 6 |
Gene Aliases: |
- |
Location: |
Chr.9:129634604-129642169 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASB6 | NM_001202403.1 | 5 | 913 | NP_001189332.1 |
| NM_017873.3 | 6 | 1000 | NP_060343.1 | |
| NM_177999.2 | 5 | 891 | NP_821066.1 | |
| AK000555.1 | 6 | 997 | BAA91250.1 | |
| AK023004.1 | 6 | 946 | BAB14355.1 | |
| AK225618.1 | 5 | 888 | ||
| AK292807.1 | 6 | 997 | ||
| AK299193.1 | 5 | 795 | ||
| BC001719.1 | 5 | 846 | AAH01719.1 | |
| BC065913.1 | 6 | 903 | AAH65913.1 | |
| BX648923.1 | 5 | 1521 | CAH10563.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469901 | Chr.9:129597897 - 129681820 on Build GRCh38 | Loss |
|
| nsv1046477 | Chr.9:129608081 - 129997996 on Build GRCh38 | Gain |
|
| nsv951784 | Chr.9:129625222 - 129655421 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map