Assay Details
Target Gene Details
Entrez Gene ID: | 10713 |
Gene Name: | ubiquitin specific peptidase 39 |
Gene Aliases: |
65K, CGI-21, HSPC332, SAD1, SNRNP65 |
Location: |
Chr.2:85602842-85649284 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP39 | NM_001256725.1 | 5 | 733 | NP_001243654.1 |
| NM_001256726.1 | 5 | 733 | NP_001243655.1 | |
| NM_001256727.1 | 5 | 618 | NP_001243656.1 | |
| NM_001256728.1 | 4 | 543 | NP_001243657.1 | |
| NM_006590.3 | 5 | 733 | NP_006581.2 | |
| NR_046347.1 | 5 | 621 | ||
| XM_006711922.1 | 5 | 601 | XP_006711985.1 | |
| XM_006711923.2 | 5 | 680 | XP_006711986.1 | |
| XM_011532487.1 | 5 | 686 | XP_011530789.1 | |
| XM_011532488.1 | 5 | 619 | XP_011530790.1 | |
| XM_017003182.1 | 5 | 690 | XP_016858671.1 | |
| AF130096.1 | 4 | 322 | AAG35521.1 | |
| AF132955.1 | 5 | 637 | AAD27730.1 | |
| AF353989.1 | 5 | 640 | AAK49524.1 | |
| AK001047.1 | 5 | 631 | ||
| AK001772.1 | ||||
| AK056334.1 | 5 | 602 | ||
| AK129872.1 | 6 | 614 | ||
| AK222852.1 | 5 | 636 | BAD96572.1 | |
| AK289451.1 | 5 | 687 | ||
| AK295257.1 | 4 | 541 | ||
| AK302236.1 | 5 | 595 | ||
| AK309485.1 | 6 | 536 | ||
| AL117416.1 | 3 | 300 | ||
| AL519827.3 | 5 | 683 | ||
| BC001384.2 | 5 | 622 | AAH01384.2 | |
| BC067273.1 | 5 | 661 | AAH67273.1 | |
| BX462623.2 | 5 | 666 | ||
| DA548921.1 |
More Information
Set Membership: |
|

Genomic Map