Assay Details
Target Gene Details
Entrez Gene ID: | 27030 |
Gene Name: | mutL homolog 3 |
Gene Aliases: |
HNPCC7 |
Location: |
Chr.14:75013764-75051532 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MLH3 | NM_001040108.1 | 13 | 7668 | NP_001035197.1 |
| NM_014381.2 | 12 | 7596 | NP_055196.2 | |
| XM_005267532.4 | 12 | 7563 | XP_005267589.1 | |
| XM_005267533.4 | 11 | 7491 | XP_005267590.1 | |
| XM_011536646.2 | 11 | 7497 | XP_011534948.1 | |
| XM_017021219.1 | 12 | 7522 | XP_016876708.1 | |
| BX090851.1 | 1 | 80 |
More Information
Additional Information:
For this assay, SNP(s) [rs77514025] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map