Assay Details
Target Gene Details
Entrez Gene ID: | 5394 |
Gene Name: | exosome component 10 |
Gene Aliases: |
PM-Scl, PM/Scl-100, PMSCL, PMSCL2, RRP6, Rrp6p, p2, p3, p4 |
Location: |
Chr.1:11066613-11099910 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EXOSC10 | NM_001001998.2 | 1 | 59 | NP_001001998.1 |
| NM_002685.3 | 1 | 59 | NP_002676.1 | |
| XM_005263475.2 | XP_005263532.2 | |||
| AB209236.1 | BAD92473.1 | |||
| AK294058.1 | ||||
| AK296559.1 | ||||
| AK308778.1 | ||||
| BC039901.1 | AAH39901.1 | |||
| BC073788.1 | AAH73788.1 | |||
| DA505395.1 | 1 | 59 | ||
| L01457.1 | AAB59352.1 | |||
| X66113.1 | CAA46904.1 |
Target Gene Details
Entrez Gene ID: | 105376736 |
Gene Name: | uncharacterized LOC105376736 |
Gene Aliases: |
- |
Location: |
Chr.1:11099675-11102105 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105376736 | NR_135061.1 | 1 | 178 | |
| NR_135062.1 | ||||
| AI342698.1 | ||||
| BE669601.1 | 1 | 179 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv955233 | Chr.1:11099444 - 11101843 on Build GRCh38 | Deletion |
|
| nsv545407 | Chr.1:11000018 - 11377969 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map