Assay Details
Target Gene Details
Entrez Gene ID: | 84913 |
Gene Name: | atonal bHLH transcription factor 8 |
Gene Aliases: |
HATH6, bHLHa21 |
Location: |
Chr.2:85753786-85791383 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATOH8 | NM_032827.6 | 2 | 1225 | NP_116216.2 |
| XM_006712122.3 | 2 | 1225 | XP_006712185.1 | |
| XM_011533139.1 | 2 | 1225 | XP_011531441.1 | |
| AF529205.1 | 2 | 821 | AAO85773.1 | |
| AK027614.1 | 2 | 1018 | BAB55233.1 | |
| AK074681.1 | 2 | 1117 | BAC11135.1 | |
| AK096049.1 | 2 | 572 | ||
| AL831857.1 | 2 | 364 | CAH56263.1 | |
| BC021207.2 | 2 | 1116 | AAH21207.1 | |
| BC094832.1 | 2 | 895 | AAH94832.1 |
More Information
Set Membership: |
|

Genomic Map