Assay Details
Target Gene Details
Entrez Gene ID: | 685 |
Gene Name: | betacellulin |
Gene Aliases: |
- |
Location: |
Chr.4:74744759-74794718 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BTC | NM_001316963.1 | 2 | 475 | NP_001303892.1 |
| NM_001729.3 | 2 | 475 | NP_001720.1 | |
| XM_011532211.1 | 2 | 456 | XP_011530513.1 | |
| XM_017008568.1 | 2 | 507 | XP_016864057.1 | |
| BI552049.1 | 2 | 291 | ||
| KU177946.1 | 2 | 114 | ||
| KU177947.1 | 2 | 114 | ||
| S55606.1 | 2 | 408 | AAB25452.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1009099 | Chr.4:74223516 - 74800996 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map