Assay Details
Target Gene Details
Entrez Gene ID: | 51131 |
Gene Name: | PHD finger protein 11 |
Gene Aliases: |
APY, BCAP, IGEL, IGER, IGHER, NY-REN-34, NYREN34 |
Location: |
Chr.13:49495610-49528992 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHF11 | NM_001040443.2 | 4 | 808 | NP_001035533.1 |
| NM_001040444.2 | 4 | 484 | NP_001035534.1 | |
| NR_135322.1 | 3 | 686 | ||
| NR_135323.1 | 5 | 620 | ||
| AB011031.1 | 4 | 457 | BAA32101.1 | |
| AF155105.1 | 4 | 427 | AAD42871.1 | |
| AK026228.1 | 4 | 448 | ||
| AK225986.1 | 5 | 570 | ||
| AK293242.1 | 4 | 808 | ||
| AK300410.1 | 4 | 448 | ||
| AL552215.3 | 4 | 616 | ||
| BC017212.2 | 4 | 437 | AAH17212.2 | |
| BX461254.2 | 3 | 316 | ||
| DA652444.1 | ||||
| DB050568.1 |
More Information
Set Membership: |
|

Genomic Map