Assay Details
Target Gene Details
Entrez Gene ID: | 4241 |
Gene Name: | melanotransferrin |
Gene Aliases: |
CD228, MAP97, MFI2, MTF1, MTf |
Location: |
Chr.3:197001740-197029816 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MELTF | NM_005929.5 | 16 | 2394 | NP_005920.2 |
| XM_006713643.3 | 16 | 2471 | XP_006713706.2 | |
| XM_011512850.2 | 16 | 2474 | XP_011511152.1 | |
| A00127.1 | CAA00012.1 |
Target Gene Details
Entrez Gene ID: | 100507057 |
Gene Name: | MFI2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:197002906-197004744 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MFI2-AS1 | NR_038285.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv822421 | Chr.3:196949827 - 197027448 on Build GRCh38 | Loss |
|
| dgv8855n54 | Chr.3:197002210 - 197003542 on Build GRCh38 | Gain |
|
| nsv1073277 | Chr.3:197002728 - 197003429 on Build GRCh38 | Deletion |
|
| esv3599293 | Chr.3:196759100 - 197444213 on Build GRCh38 | Gain |
|
| dgv8857n54 | Chr.3:197002559 - 197003542 on Build GRCh38 | Loss |
|
| nsv517987 | Chr.3:196990442 - 197023970 on Build GRCh38 | Loss |
|
| nsv1144060 | Chr.3:197002829 - 197003429 on Build GRCh38 | Deletion |
|
| nsv1003135 | Chr.3:196621850 - 197012082 on Build GRCh38 | Gain |
|
| esv29143 | Chr.3:197002152 - 197003909 on Build GRCh38 | Loss |
|
| esv2763775 | Chr.3:196727069 - 197015419 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map