Assay Details
Target Gene Details
Entrez Gene ID: | 10313 |
Gene Name: | reticulon 3 |
Gene Aliases: |
ASYIP, HAP, NSPL2, NSPLII, RTN3-A1 |
Location: |
Chr.11:63681315-63759891 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RTN3 | NM_001265589.1 | 3 | 1175 | NP_001252518.1 |
| NM_001265590.1 | 2 | 839 | NP_001252519.1 | |
| NM_001265591.1 | NP_001252520.1 | |||
| NM_006054.3 | NP_006045.1 | |||
| NM_201428.2 | 2 | 1118 | NP_958831.1 | |
| NM_201429.2 | NP_958832.1 | |||
| NM_201430.2 | NP_958833.1 | |||
| NR_049750.1 | ||||
| NR_049751.1 | ||||
| XM_011544730.2 | 3 | 1265 | XP_011543032.1 | |
| XM_011544731.2 | XP_011543033.1 | |||
| XM_017017091.1 | 3 | 1265 | XP_016872580.1 | |
| XM_017017092.1 | 3 | 1265 | XP_016872581.1 | |
| AB209771.1 | BAD93008.1 | |||
| AF059524.1 | AAC99319.1 | |||
| AF119297.1 | AAD26810.1 | |||
| AK075412.1 | ||||
| AK127079.1 | 2 | 814 | ||
| AK222898.1 | BAD96618.1 | |||
| AK295361.1 | ||||
| AK295534.1 | ||||
| AK295758.1 | 2 | 767 | ||
| AK297529.1 | ||||
| AK297533.1 | ||||
| AK304100.1 | 3 | 1125 | ||
| AK307811.1 | 2 | 1068 | ||
| AK308083.1 | ||||
| AK309766.1 | 2 | 793 | ||
| AY427821.2 | 2 | 1085 | AAR02474.1 | |
| AY750848.1 | 3 | 1142 | AAU81930.1 | |
| BC000634.1 | AAH00634.1 | |||
| BC010556.1 | AAH10556.1 | |||
| BC011394.1 | AAH11394.1 | |||
| BC022993.1 | AAH22993.1 | |||
| BC100822.2 | AAI00823.1 | |||
| BC100823.2 | AAI00824.1 | |||
| BC105981.1 | AAI05982.1 | |||
| BC105982.1 | AAI05983.1 | |||
| BC118550.1 | ||||
| BC118628.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832183 | Chr.11:63649206 - 63797093 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map