Assay Details
Target Gene Details
Entrez Gene ID: | 7516 |
Gene Name: | X-ray repair cross complementing 2 |
Gene Aliases: |
FANCU |
Location: |
Chr.7:152646498-152676165 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| XRCC2 | NM_005431.1 | NP_005422.1 | ||
| AF035587.1 | AAC05369.1 | |||
| AK313607.1 | ||||
| BC042137.1 | AAH42137.1 | |||
| Y08837.1 | CAA70065.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1017565 | Chr.7:152450845 - 152758187 on Build GRCh38 | Gain |
|
| nsv831193 | Chr.7:152656459 - 152895593 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map