Assay Details
Target Gene Details
Entrez Gene ID: | 55119 |
Gene Name: | pre-mRNA processing factor 38B |
Gene Aliases: |
NET1 |
Location: |
Chr.1:108692292-108701803 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRPF38B | NM_018061.2 | 6 | 3459 | NP_060531.2 |
| NR_037185.1 | 7 | 3509 | ||
| XM_017001596.1 | 6 | 2964 | XP_016857085.1 | |
| XM_017001597.1 | 7 | 3008 | XP_016857086.1 | |
| XM_017001598.1 | 7 | 4551 | XP_016857087.1 | |
| AK001192.1 | BAA91546.1 | |||
| AL833950.1 | 7 | 3481 | CAH10702.1 |
More Information
Set Membership: |
|

Genomic Map