Assay Details
Target Gene Details
Entrez Gene ID: | 5923 |
Gene Name: | Ras protein specific guanine nucleotide releasing factor 1 |
Gene Aliases: |
CDC25, CDC25L, GNRP, GRF1, GRF55, H-GRF55, PP13187, ras-GRF1 |
Location: |
Chr.15:78959947-79090873 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 28 - Intron 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RASGRF1 | NM_001145648.1 | NP_001139120.1 | ||
| NM_002891.4 | NP_002882.3 | |||
| NM_153815.2 | NP_722522.1 | |||
| XM_011521867.2 | XP_011520169.1 | |||
| XM_017022455.1 | XP_016877944.1 | |||
| XM_017022456.1 | XP_016877945.1 | |||
| XM_017022459.1 | XP_016877948.1 | |||
| AK226101.1 | ||||
| L26584.1 | AAA58417.1 | |||
| M91815.1 | ||||
| S62035.1 | AAB26881.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33573 | Chr.15:78761142 - 79001539 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map