Assay Details
Target Gene Details
Entrez Gene ID: | 7701 |
Gene Name: | zinc finger protein 142 |
Gene Aliases: |
pHZ-49 |
Location: |
Chr.2:218637592-218659632 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF142 | NM_001105537.2 | 8 | 3863 | NP_001099007.1 |
| NR_073599.1 | 9 | 4172 | ||
| NR_073600.1 | 9 | 4005 | ||
| XM_006712730.3 | 9 | 4538 | XP_006712793.1 | |
| XM_011511784.2 | 9 | 4633 | XP_011510086.1 | |
| XM_011511785.2 | 9 | 4558 | XP_011510087.1 | |
| XM_011511786.2 | 9 | 4463 | XP_011510088.1 | |
| XM_011511787.2 | 8 | 4458 | XP_011510089.1 | |
| XM_011511788.2 | 8 | 4363 | XP_011510090.1 | |
| XM_011511789.2 | 8 | 4031 | XP_011510091.1 | |
| XM_017004871.1 | 8 | 4614 | XP_016860360.1 | |
| XM_017004872.1 | 7 | 4014 | XP_016860361.1 | |
| AK095027.1 | ||||
| D87073.1 | 8 | 3861 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3594346 | Chr.2:218640692 - 218643860 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map